SP Bel-Art Glove box, portable
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H500282000 H500282001
BELAH500282000EA 4730 EUR
BELAH500282000 BELAH500282001
SP Bel-Art Glove box, portable
Handschoenenkasten en -zakken
This lightweight, self-contained portable isolation cabinet made of LDPE protects the operator and the external environment.
  • Large acrylic window with 6" circular glove ports permits safe viewing of operations
  • 8” diameter side port allows easy insertion of material and equipment
  • Large size, 22" long (20" installed) neoprene gloves (included for some models only) provide good protection against gas, liquid or solid penetration
  • Gloves are secured to the 6" circular port holes with clamping rings (included for some models only)
  • Body molded from LDPE with round corners throughout for easy cleaning
Available with or without gas ports. Ideal for quick set-up anywhere.
Glove port Ø: 152 mm
Bestelinformatie: The glove box is available in three different configurations: Glove box only, glove box with neoprene gloves and clamping rings, and glove box with two gas ports, neoprene gloves, and clamping rings.