Scoops, large volume, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H367590000 H367580001
BELAH367590000EA 162 EUR
BELAH367590000 BELAH367580001
Scoops, large volume, SP Bel-Art
Scheppen
PE, white. These cylindrical shaped sampling scoops are designed for retrieving samples from a larger source. Ideal for scooping dry powders, granules and crystals.
  • Closed back, non spill shape
  • Ergonomically angled handle