Filter paper discs for Buchner table-top funnels, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H146320010 H146320018 H146320024
BELAH146320010EA 299 EUR
BELAH146320010 BELAH146320018 BELAH146320024
Filter paper discs for Buchner table-top funnels, SP Bel-Art
Filters Filterpapier Kwantitatief filterpapier
These heavy bond grade filter paper discs are custom-sized to fit table-top Buchner funnels and have a thickness of 0,30 mm.
  • Made from creped white cellulose to improve porosity
  • Flow rate: 90 ml per minute
  • Wet burst strength is greater than 68 KPa; able to retain particles as small as 3 microns