Instrument tray with cover, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F161910000 F161880000
BELAF161910000EA 111 EUR
BELAF161910000 BELAF161880000
Instrument tray with cover, SP Bel-Art
Reservoirs/Schalen/Potten Instrumentenschalen
Made of PP, translucent, with inset covers.
  • Smooth and rounded corners provide easy cleaning and sterilisation
  • Cover taper gracefully outward toward the top
  • Steam autoclavable at 121 ºC