Tubing, rubber, porous
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3301025 3301226 3301530 3302235
DENE3301025EA 725 EUR
DENE3301025 DENE3301226 DENE3301530 DENE3302235
Tubing, rubber, porous
Slangen Transfer Tubing
Cellular rubber, black.
  • Flame resistant
  • Covered with special skin
  • Use for isolation
Synthetic natural rubber. Highly flexible and closed insulation material with low thermal conductivity.