Utility trays, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F163001520 F163001216 F163002020 F163001015 F163002226 F163001818
216-2742EA 418 EUR
216-2742 BELAF163001216 BELAF163002020 BELAF163001015 BELAF163002226 BELAF163001818
Utility trays, SP Bel-Art
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
PP, white.
  • Straight sided
  • Temperature resistant up to 135 ºC
  • Easy to clean