Racks, side up, for chest and upright freezers
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TE21390 TE21391 TE21392 TE21394 TE21395 TE21396
TENATE21390EA 73.69 EUR
TENATE21390 TENATE21391 TENATE21392 TENATE21394 TENATE21395 TENATE21396
Racks, side up, for chest and upright freezers
Rekken Rekken voor cryogene dozen
The side up rack both fits into chest and upright freezers with a purpose to fill out the space that is left in freezers while using a regular rack.
  • Made of stainless steel
  • Store up to 13% more samples
  • With a locking rod to secure the cryoboxes