Tubing, PE
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3550228 3550305 3550406 3550608 3550709 3551012 3550507 3550811 3550911 3551014 3550204 3551216 3550304 3550609 3551215 3550118 3551535 3550405 3550710 3550810 3550913 3551013 3551214
228-3134EA 1.7 EUR
228-3134 228-3140 228-3142 228-3146 228-3152 228-3168 DENE3550507 DENE3550811 DENE3550911 DENE3551014 DENE3550204 228-3000 DENE3550304 DENE3550609 DENE3551215 DENE3550118 DENE3551535 DENE3550405 DENE3550710 DENE3550810 DENE3550913 DENE3551013 DENE3551214
Tubing, PE
Slangen Transfer Tubing
PE, translucent
  • Odorless and tasteless
  • Resistant to acids, bases, alcohols, and salt solutions up to temperature of 80 °C
  • Flexible