Microplate racks for upright freezers
Leverancier: Tenak
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TENATE27480EA 121.51 EUR
TENATE27480 TENATE27482 TENATE27560 TENATE27484 TENATE27440 TENATE27442 TENATE27564 TENATE27520 TENATE27488 TENATE27400 TENATE27522 TENATE27402 TENATE27600 TENATE27404 TENATE27604 TENATE27406 TENATE27606 479-1168 TENATE27490 479-1167 TENATE27570 TENATE27450 TENATE27452 TENATE27530 TENATE27574 TENATE27410 TENATE27576 TENATE27532 TENATE27412 TENATE27610 TENATE27456 479-1164 TENATE27458 TENATE27414 479-1163 TENATE27612 TENATE27536 479-1166 TENATE27614 479-1165 TENATE27416 TENATE27580 TENATE27460 TENATE27582 TENATE27462 TENATE27540 TENATE27584 479-2183 TENATE27420 TENATE27586 TENATE27542 TENATE27422 479-2185 TENATE27466 TENATE27500 TENATE27544 479-2184 TENATE27468 TENATE27424 479-2187 479-2186 TENATE27502 479-2189 TENATE27426 479-2188 TENATE27548 TENATE27506 TENATE27508 479-2190 TENATE27590 TENATE27470 TENATE27592 TENATE27594 TENATE27550 TENATE27430 TENATE27596 TENATE27552 TENATE27432 TENATE27598 TENATE27510 TENATE27434 TENATE27558 TENATE27514
Microplate racks for upright freezers
Rekken Rekken voor microplaten
Complete range of racks for microtiter plates for use in upright freezers, made of stainless steel. The racks can either be used as one plate per shelf or the plates can be stacked.