Swabs in tubes
Leverancier: HERENZ HEINZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1020001 1020002 1020003 1020025
HERE1020001EA 135 EUR
HERE1020001 HERE1020002 HERE1020003 HERE1020025
Swabs in tubes
Wattenstaafjes Wattenstaafjes in buisjes
Sterile dry swabs in tubes, without transport medium.
  • Handle is made of PS, aluminium or wood
  • Head is made of viscose/rayon or cotton
  • Unbreakable PP tube (approx. 150×12 mm)
With label on the tube and stopper.
Verpakking: Available loose or individually packed in a peel pack.