Butter trier
Leverancier: GERBER FUNKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3131 3130
300-0147EA 161 EUR
300-0147 300-0146
Butter trier
Monsternemers Monsternemers voor vaste stoffen
Chrome-nickel steel. Developed to the test the firmness and consistency of butter. For use in butter making industries.
  • With metal handle for improved hygiene
  • Two different sizes available