Tubing, butyl
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3220610 3220812 3221014 3220614
DENE3220610EA 11.4 EUR
DENE3220610 DENE3220812 DENE3221014 DENE3220614
Tubing, butyl
Slangen Transfer Tubing
Black butyl rubber tubing.
Largely impermeable to gas, without reinforcement.
Temperature range up to +100 °C.
Order Now
Specificaties
Environmentally Preferable