Tubing, latex rubber
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3050610 3050711 3050305 3051014 3050608 3050508 3050206 3050812 3050204 3050307 3050406 3050507 3051016 3051217 3050306 3050509 3050710 3050913 3050914 3050407 3050408 3050609
228-3352EA 77.4 EUR
228-3352 228-3357 DENE3050305 DENE3051014 DENE3050608 DENE3050508 DENE3050206 DENE3050812 DENE3050204 DENE3050307 DENE3050406 DENE3050507 DENE3051016 DENE3051217 DENE3050306 DENE3050509 DENE3050710 DENE3050913 DENE3050914 DENE3050407 DENE3050408 DENE3050609
Tubing, latex rubber
Slangen Transfer Tubing
Natural Rubber (NR), translucent.
  • Sterilisable at +120 °C
  • Hardness: 40 Shore A