Ureum, EMPROVE® api BP, Ph. Eur., USP suitable for use as an active pharmaceutical ingredient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
1.08432.5000 1.08432.9029
1.08432.5000EA 1290 EUR
1.08432.5000 1.08432.9029
Ureum, EMPROVE® api BP, Ph. Eur., USP suitable for use as an active pharmaceutical ingredient, SAFC®
Ureum
Formule: NH₂CONH₂
MW: 60.06 g/mol
Kookpunt: 197
Smeltpunt: 132.7
Dichtheid: 1.34 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
Bereik: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM