2,4,6-Trimethylphenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01622.1 SB01622.10 SB01622.50
MAYBSB01622.1EA 0 EUR
MAYBSB01622.1 MAYBSB01622.10 MAYBSB01622.50
2,4,6-Trimethylphenyl isothiocyanate
2,4,6-Trimethylphenyl isothiocyanate
Formule: C₁₀H₁₁NS
Kookpunt: 75 °C (0,3 mmHg)
Smeltpunt: 64 °C
MDL Nummer: MFCD00022055
CAS nummer: 6095-82-5
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR