N-(2,4-Dichlorophenyl)-2-chloroacetamide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-Chloro-N-(2,4-dichlorophenyl)acetamide, N-(Chloroacetyl)-2,4-dichloroaniline
KM06507.250 KM06507.1
MAYBKM06507.250EA 0 EUR
MAYBKM06507.250 MAYBKM06507.1
N-(2,4-Dichlorophenyl)-2-chloroacetamide
N-(2,4-Dichlorophenyl)-2-chloroacetamide
Formule: C₈H₆Cl₃NO
MDL Nummer: MFCD00018888
CAS nummer: 6974-56-7