Tetraethylurea ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
1,1,3,3-Tetraethylurea
L09458.06
L09458.14
L09458.06EA
28.8
EUR
L09458.06
L09458.14
Tetraethylurea ≥99%
Tetraethylurea
Purity: 99%
CAS No.: 1187-03-7
Molecular Formula: (CH3CH2)2NCON(CH2CH3)2
Formula Weight: 172.27
Flash Point: 81°(177°F)
Refractive Index: 1.4470
Boiling Point: 212-214°
Beilstein Reference: 1763126
MDL No.: MFCD00042881
Formule:
C₉H₂₀N₂O MW: 172.27 g/mol Kookpunt: 212…214 °C Dichtheid: 0.910 Vlampunt: 81 °C (177 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00042881 CAS nummer: 1187-03-7 EINECS: 214-696-0 |