4-Isopropylbenzyl chloride 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-(Chloromethyl)-4-isopropylbenzene
L11839.06
L11839.03
L11839.06EA
53.2
EUR
L11839.06
L11839.03
4-Isopropylbenzyl chloride 95%
4-Isopropylbenzyl chloride
Purity: 95%
CAS No.: 2051-18-5
Molecular Formula: C10H13Cl
Formula Weight: 168.67
Flash Point: 104°(219°F)
Refractive Index: 1.5230
Odour: Pungent
Boiling Point: 119-120°/35mm
Beilstein Reference: 907207
UN No.: 3265
MDL No.: MFCD00018885
Formule:
C₁₀H₁₃Cl Kookpunt: 119…120 °C (35 mmHg) Dichtheid: 1,020 Vlampunt: 104 °C (219 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00018885 CAS nummer: 2051-18-5 EINECS: 218-114-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...