Potassium tetrachloroaurate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Gold potassium chloride, Potassium tetrachloroaurate(III), Potassium gold(III) chloride
316120010 316120050
ACRO316120010EA 167 EUR
ACRO316120010 ACRO316120050
Potassium tetrachloroaurate 98%
Potassium tetrachloroaurate
Formule: AuCl₄K
MW: 377.88 g/mol
Smeltpunt: 357 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00043088
CAS nummer: 13682-61-6
EINECS: 237-190-1
UN: 3260
Order Now
Specification Test Results
Appearance Yellow to light orange Crystalline powder
Assay ≥97.5 %