Dodecylphosphonic acid 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
H26259.03
H26259.09
H26259.03EA
40.8
EUR
H26259.03
H26259.09
Dodecylphosphonic acid 95%
Dodecylphosphonic acid
Purity: 95%
CAS No.: 5137-70-2
Molecular Formula: CH3(CH2)11P(O)(OH)2
Formula Weight: 250.31
Melting Point: 92-96°
UN No.: 3261
MDL No.: MFCD00015832
Formule:
C₁₂H₂₇O₃P Smeltpunt: 92…96 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00015832 CAS nummer: 5137-70-2 EINECS: 225-897-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...