Manganese(II) perchlorate hexahydrate ≥99.995% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
44318.09 44318.18
44318.09EA 91.7 EUR
44318.09 44318.18
Manganese(II) perchlorate hexahydrate ≥99.995% (metals basis)
Manganese(II) perchlorate hexahydrate
Purity: 99.995% CAS No.: 15364-94-0 Molecular Formula: Mn(ClO4)2.6H2O Formula Weight: 361.93 (253.84anhy) Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 1481 MDL No.: MFCD00150255
Formule: Cl₂MnO₈·6H₂O
MW: 361.931 g/mol
Smeltpunt: 165 °C. (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150255
CAS nummer: 15364-94-0
EINECS: 237-390-9
UN: 1481
ADR: 5.1 II