3-Nitro-L-tyrosine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4026245.0025 4026245.0100
BACE4026245.0025EA 0 EUR
BACE4026245.0025 BACE4026245.0100
3-Nitro-L-tyrosine
3-Nitro-L-tyrosine
Formule: C₉H₁₀N₂O₅
MDL Nummer: MFCD00007123
CAS nummer: 621-44-3
EINECS: 210-688-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR