4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
L00982.06 L00982.14
L00982.06EA 56.6 EUR
L00982.06 L00982.14
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole ≥99%
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Purity: 99+% CAS No.: 1750-12-5 Molecular Formula: C2H6N6S Formula Weight: 146.17 Melting Point: ca 220° dec. Beilstein Reference: 134867 UN No.: 1325 MDL No.: MFCD00003098
Formule: C₂H₆N₆S
MW: 146.17 g/mol
Smeltpunt: ∼ 220 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003098
CAS nummer: 1750-12-5
EINECS: 217-135-8
UN: 1325
ADR: 4.1 III