Triethyl-1,1,2-ethanetricarboxylate 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Triethyl ethane-1,1,2-tricarboxylate
139610250 139611000
ACRO139610250EA 35 EUR
ACRO139610250 ACRO139611000
Triethyl-1,1,2-ethanetricarboxylate 99%
Triethyl-1,1,2-ethanetricarboxylate
Formule: C₁₁H₁₈O₆
MW: 246.26 g/mol
Kookpunt: 99 °C (.5 mmHg)
Dichtheid: 1.07 g/cm³
Vlampunt: >112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009154
CAS nummer: 7459-46-3
EINECS: 231-235-9
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥98.5 %