3,4',5-Trimethoxybenzophenone
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10004185-10 10004185-100 10004185-50 10004185-25
CAYM10004185-10EA 52.6 EUR
CAYM10004185-10 CAYM10004185-100 CAYM10004185-50 CAYM10004185-25
3,4',5-Trimethoxybenzophenone
3,4',5-Trimethoxybenzophenone
Formule: C₁₆H₁₆O₄
CAS nummer: 94709-12-3