(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L09031.06
L09031.14
L09031.06EA
24
EUR
L09031.06
L09031.14
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98%
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione
Purity: 98+%
CAS No.: 4511-42-6
Molecular Formula: C6H8O4
Formula Weight: 144.13
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4475
Melting Point: 95-99°
Boiling Point: 255°
Beilstein Reference: 82124
Optical Rotation: +265° (c=1 in chloroform)
MDL No.: MFCD00070594
Formule:
C₆H₈O₄ Kookpunt: 255 °C Smeltpunt: 95…99 °C |
MDL Nummer:
MFCD00070594 CAS nummer: 4511-42-6 EINECS: 224-832-0 |