S-Methylisothiouronium sulphate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate
127891000 127895000
ACRO127891000EA 28.8 EUR
ACRO127891000 ACRO127895000
S-Methylisothiouronium sulphate 98%
S-Methylisothiouronium sulphate
Formule: C₄H₁₂N₄S₂·H₂SO₄
MW: 278.38 g/mol
Smeltpunt: 240…241 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013055
CAS nummer: 867-44-7
EINECS: 212-759-7
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
HPLC ≥97.5 %