Inlet solvent filters, Bottom-of-the-Bottle™, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-426 A-437 A-438 A-445 A-446 A-550 A-551
UPCHA-426EA 67.1 EUR
UPCHA-426 UPCHA-437 554-1977 UPCHA-445 554-3041 554-3046 UPCHA-551
Inlet solvent filters, Bottom-of-the-Bottle™, Upchurch Scientific®
Filters Filters voor vloeistofverwerking
Stainless steel, PEEK or UHMWPE
  • Stainless steel filters draw solvents from within ¹/₈" of the bottom of the bottle
  • PEEK filters draw solvents from within 0,080" (2,0 mm) of the bottom of the bottle
  • UHMWPE filters draw solvents from within 0,10" (2,5 mm) of the bottom of the bottle