Biocompatible precolumn filters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-355 A-355-01 A-355-02 A-356
554-2212EA 165 EUR
554-2212 554-3033 554-3034 554-2213
Biocompatible precolumn filters, Upchurch Scientific®
Filters Voorkolom filters en frits
Precolumn filters, PEEK bodies and PCTFE-surrounded PEEK frits.
  • Complete with 0,50 mm Ø thru-holes and 8° distribution cones for minimal band spreading and mixing
  • Designed for use with ¹/₁₆" Ø tubing
  • Pressure rated to 5000 psi (345 bar)