VWR®, TE (Tris-EDTA) buffer, pH 8,0 for biotechnology
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E112-100MLEA
49.3
EUR
E112-100ML
E112-500ML
VWR®, TE (Tris-EDTA) buffer, pH 8,0 for biotechnology
Buffers
Kloonbuffers
Ready to use Tris-EDTA buffer.
- Composition: 10 mM Tris, 1 mM EDTA
- pH 8,0