Temperature probes, K-type, with SMP-connector, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1860-20 P1860-15 P1860-25
BOHLP1860-20EA 201 EUR
BOHLP1860-20 BOHLP1860-15 BOHLP1860-25
Temperature probes, K-type, with SMP-connector, BOLA
Temperatuursondes
Thermocouple in a PTFE encapsulated stainless steel tube that is ideal for temperature measurement in aggressive liquids and the cable provides flexible connection from measuring device to medium.
  • With a pointed tip, collar ring Ø 12 mm and a green PTFE coated cable and SMP connector
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance