VWR® UltraPure, PBS and TBS wash buffers
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0788-2PKEA
391
EUR
0788-2PK
E703-1L
E703-500ML
J640-4L
VWR® UltraPure, PBS and TBS wash buffers
Buffers
Buffers voor elektroforese
Wasbuffers
Wash buffers ideal for use in applications such as Western blotting and ELISA. Prevents high background by effectively removing unbound reagents.