VWR®, SSC buffer, 20X, ultra-pure grade
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
0804-4LEA 225 EUR
0804-4L
VWR®, SSC buffer, 20X, ultra-pure grade
Buffers Buffers voor elektroforese Hybridisatiebuffers
SSC buffer is a hybridisation buffer for Northern and Southern blots.
Leveringsinformatie: Supplied as either 20X liquid concentrate or Ready-Pack™ that prepares 1 litre of 20X concentrate.