VWR® UltraPure, Tris-acetate-EDTA (TAE) buffer
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K915-1.6LEA
192
EUR
K915-1.6L
K915-4L
K915-5L
VWR® UltraPure, Tris-acetate-EDTA (TAE) buffer
Buffers
Buffers voor elektroforese
Running-buffers
TAE is an extensively used buffer for agarose gel electrophoresis applications requiring high resolution and separation of high molecular weight, double-stranded DNA. TAE buffer is more compatible with in-gel manipulations and band recovery procedures than TBE buffer.
- Ideal for DNA recovery and in-gel manipulations
- Low ionic strength
- Better resolution of large DNA fragments (>12 kb DNA)
- Convenient packaging options minimise weighing and mixing
Composition of (1X) TAE buffer: 0,04 M Tris-acetate; 0,001 M EDTA; final pH 8,0.