Tubing, Viton (FPM)
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3200609B 3200913 3200811 3201012 3201216 3200608 3200305 3201014 3200406 3200507 3200610 3200709 3200711 3200812 3200912 3200609 3200407 3200508 3200408 3200710 3200810
228-2351EA 32.9 EUR
228-2351 228-4323 228-4324 DENE3201012 DENE3201216 DENE3200608 DENE3200305 DENE3201014 DENE3200406 DENE3200507 DENE3200610 DENE3200709 DENE3200711 DENE3200812 DENE3200912 DENE3200609 DENE3200407 DENE3200508 DENE3200408 DENE3200710 DENE3200810
Tubing, Viton (FPM)
Slangen Transfer Tubing
Fluororubber (FPM), black.
  • Temperatur resistance: −10 to +200 °C
  • Hardness: 75 Shore A
  • Dimension tolerances according to DIN ISO 3302-1 E2
Good resistance against ozone and climatic exposure as well as gasoline and mineral oil.