Dialysis tubing funnel, Fillpour™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F148480001 F148480000
229-0002EA 179 EUR
229-0002 BELAF148480000
Dialysis tubing funnel, Fillpour™, SP Bel-Art
Trechters Filtertrechters Filtertrechters
Dialysis tubing is cut to length and knotted at the end. The other end is slipped onto the tube and is held with silicone rubber loops or a rubber band. Three tube steps fit various sizes of dialysis tubing, and a triple notched universal hanger supports the funnel on the edge of any vessel.
  • Reduces difficulties in handling narrow dialysis bags
  • Captive cap with writing area
  • Steam autoclavable at 121 °C