Filter thimbles with 38 mm flange for dietary fibre extraction, VitraPOR®
Leverancier: ROBU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VitraPOR®
ROBU20684EA 152 EUR
ROBU20684 ROBU20682 ROBU20683 ROBU20680 ROBU20681
Filter thimbles with 38 mm flange for dietary fibre extraction, VitraPOR®
Extractors Extractiehulzen
Borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Filter-Thimbles, CFE, with exact glass flange fits many current systems.