Magnetic stirring bars, PTFE, polygonal, without pivot ring, Spinbar®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F371200015 F371200020 F371200025 F371200030 F371200040 F371200050 F371200060 F371200080 F371200010 F371200038
442-4228EA 21.8 EUR
442-4228 442-4229 442-4230 BELAF371200030 BELAF371200040 BELAF371200050 BELAF371200060 BELAF371200080 442-4227 BELAF371200038
Magnetic stirring bars, PTFE, polygonal, without pivot ring, Spinbar®, SP Bel-Art
Roerders Magnetische roerstaven
Polygonal PTFE encapsulated stirring bars.
  • Multifaceted surfaces that add turbulence relative to similar sized smooth cylindrical stirring bars