Drying tubes with tube fittings, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F199600000 F199610000 F199620000
BELAF199600000EA 67.4 EUR
BELAF199600000 BELAF199610000 BELAF199620000
Drying tubes with tube fittings, SP Bel-Art
Buisjes Droogbuizen
Unbreakable polyethylene straight tubes.
  • Good chemical resistance
  • Godd temperature resistance
  • Drying agent no included
Can be filled with desiccant of your choice. Each tube is supplied with two polypropylene fittings which accept 6,4 to 9,5 mm int.Ø tubing.