VWR®, DNA ladder, 100 bp
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
K180-250ULEA 176 EUR
K180-250UL
VWR®, DNA ladder, 100 bp
Reagentia voor elektroforese Molecuulgewichtmerkers
Ladder with 13 fragments ranging from 100 to 3000 bp for easy band identification.
Also available pre-mixed with loading dye.