VWR®, Protease inhibitor cocktail, mammalian
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
M250-1MLEA 104 EUR
M250-1ML
VWR®, Protease inhibitor cocktail, mammalian
Celcultuursupplementen en -additieven
Contains protease inhibitors for mammalian cell use: AEBSF, Aprotinin, E-64, Bestatin, Leupeptin and Pepstatin