Sample bags, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H131780812 H131761016 F131760816 F131761220
BELAH131780812EA 103 EUR
BELAH131780812 BELAH131761016 BELAF131760816 BELAF131761220
Sample bags, SP Bel-Art
Zakken Monsterzakken
These utility reusable, transparent PE bags have variety of applications such as they are ideal for shipping, storage and are especially useful as sample bags.
  • Not autoclavable
  • Latex-free
  • Available in two weights
Certificaten: FDA approved.