Contact plate racks, Poxygrid®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F189790002 F189790003
BELAF189790002EA 170 EUR
BELAF189790002 BELAF189790003
Contact plate racks, Poxygrid®, SP Bel-Art
Rekken Dispensers voor petrischalen
Epoxy coated steel wire. Holds 10 or 30 contact plates with lids arranged in one or three stacks of ten plates.
  • Prevents accidents during transport
  • Chemical and corrosion resistant
  • Compact size maximises incubator space and won't crowd bench top
W×D×H: 82×82×140 cm (one stack)
W×D×H: 246×82×140 mm (three stacks)