Salt tester for dairy products
Leverancier: GERBER FUNKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4760 4770
614-0194EA 109 EUR
614-0194 614-0195
Salt tester for dairy products
Testsystemen voor voeding en drank
Titration method with Dr Schilling pattern burettes and brown PE bottle.