Sediment testers for milk
Leverancier: GERBER FUNKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4800 4810
511-0076EA 387 EUR
511-0076 614-0196
Sediment testers for milk
Filterhouders
SEDILAB: Manual sediment tester. With clamp for attaching to tables.
SEDILAB-E: Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk. Splash-proof design. Approx. 800 samples per hour. Sharply defined sediment images.
Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk.