Industrial label makers, benchtop, GlobalMark®
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
250353 250354
BRDY250353EA 4695 EUR
BRDY250353 116-0268
Industrial label makers, benchtop, GlobalMark®
Printers Label Printers
The GlobalMark® label printer is a rugged, easy to use, industry-ready printer designed for large volumes of print. It can be used to print custom labels, tags, stickers, badges and signs as needed.
  • Versatile print results with an industry-standard
  • Over 700 built-in symbols
  • Prints on labels and tapes having width of 12,70 to 101 mm
Advanced touch screen technology with the Microsoft Windows® CE operating system makes these printers incredibly fast and easy to create professional labels. This can be used either as standalone unit or connected to PC using MarkWare™ facility identification software.