Desiccator cabinets, Secador®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F420721009 F420731002 F420741002 F420731009 F420710002
BELAF420721009EA 1590 EUR
BELAF420721009 BELAF420731002 BELAF420741002 BELAF420731009 BELAF420710002
Desiccator cabinets, Secador®, SP Bel-Art
Exsiccatoren Exsiccatorkasten
The attractive design and sturdy construction of the Secador® range of desiccator cabinets ensures easy and reliable storage of moisture and UV sensitive products. Durastar® co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack.
  • Large doors maximise access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Rear mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
  • Perforated shelves facilitate circulation of dry air and have space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located on the front of the door that opens to the left
Amber coloured units reduce visible light penetration by over 50% so are the perfect choice for storing light sensitive materials such as reagents, analytical standards and investigational compounds. 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
Units have a space saving 341×414 mm (W×D) footprint and can be stacked up to 3 units high.
Bestelinformatie: Replacement shelves are available on request.
Opgelet: These desiccator cabinets are not suitable for use with vacuum.