Aspirator bottles, vacuum, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F199170001 F199170002
BELAF199170001EA 544 EUR
BELAF199170001 BELAF199170002
Aspirator bottles, vacuum, SP Bel-Art
Flessen Doseerflessen
Polypropylene, with epoxy coated steel frame and screw cap.
  • Chemical resistant
  • Corrosion resistant
  • Eopxy coated steel frame saves workspace
Complete aspirator bottle includes vaccum trap, safety vent on screw cap, flexible collection tubing (1/4"×3/8") and UV inhibitor molded collection bottle.
Note: Disposable pipette tip, pasteur pipette needs to be connect to collection tubing for vaccum aspiration from petri dishes, microtitre plates, centrifuge tubes and flasks.