Transport tubes, selfstanding, non sterile
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T550-10ATPR T550-10AT T552-10ATTP T552-12ATTP T552-30ATTP T552-5ATTP T552-7ATTP
212-9609EA 252 EUR
212-9609 SIMPT550-10AT SIMPT552-10ATTP SIMPT552-12ATTP SIMPT552-30ATTP SIMPT552-5ATTP SIMPT552-7ATTP
Transport tubes, selfstanding, non sterile
Monsternamepotjes Containers voor algemeen gebruik
Tubes made of non-toxic polypropylene, caps made of polyethylene. Designed for storage and transportation of biological material.
  • Good chemical resistance
  • Good temperature resistance
  • All graduated tubes are in 0,5 ml increments
Tubes provide strength and clarity and exhibit some unique design features. Five sizes are available from 5 to 30 ml. The 212-9609 tube has a white marking area to make sample identification more convenient. They have external threads to provide a smooth and uniform inner surface. A perfect leakproof seal is obtained by the use of a specially designed flexible sealing lip inside the polyethylene closures. Cap also feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn.
Bestelinformatie: Screw caps have to be ordered separately.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904