Hygromycin B 50 mg/ml in aqueous solution
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
CP12.2 CP12.1
ROTHCP12.2EA 215 EUR
ROTHCP12.2 ROTHCP12.1
Hygromycin B 50 mg/ml in aqueous solution
Hygromycin B
Formule: C₂₀H₃₇N₃O₁₃
MDL Nummer: MFCD06795479
CAS nummer: 31282-04-9
ADR: 6.1,II